Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylacetone 98.0+%, TCI America™
SDP

Catalog No. F053525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F053525G 25 g
F05355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F053525G Supplier TCI America Supplier No. F053525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 459-03-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00000362
InChI Key ZUEKIIWSVFBTCM-UHFFFAOYSA-N
Synonym 4-fluorophenylacetone, 4-fluorophenyl acetone, 1-4-fluorophenyl propan-2-one, 1-4-fluorophenyl acetone, 1-4-fluorophenyl-2-propanone, 2-propanone, 1-4-fluorophenyl, p-fluorophenyl acetone, 1-acetonyl-4-fluorobenzene, 1-4-fluoro-phenyl-propan-2-one
PubChem CID 521187
IUPAC Name 1-(4-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(F)C=C1

Specifications

Boiling Point 130°C
Quantity 25 g
Formula Weight 152.17
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Fluorophenylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.