Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylacetonitrile, 98+%

Catalog No. AAA1429014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1429014 25 g
AAA1429006 5 g
AAA1429022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1429014 Supplier Thermo Scientific Chemicals Supplier No. A1429014
Add to Cart

CAS: 459-22-3 | C8H6FN | 135.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-22-3
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00001917
InChI Key JHQBLYITVCBGTO-UHFFFAOYSA-N
Synonym 4-fluorophenylacetonitrile, 4-fluorobenzyl cyanide, p-fluorobenzyl cyanide, benzeneacetonitrile, 4-fluoro, 4-fluorobenzylcyanide, 4-fluorobenzeneacetonitrile, p-fluorophenylacetonitrile, 2-4-fluorophenyl acetonitrile, 4-fluorobenzylnitrile, acetonitrile, p-fluorophenyl
PubChem CID 68016
IUPAC Name 2-(4-fluorophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)F

Specifications

Density 1.13
Boiling Point 238°C
Flash Point 108°C (226°F)
Refractive Index 1.5002
Quantity 25 g
UN Number UN3276
Beilstein 1907764
Formula Weight 135.14
Percent Purity ≥98%
Chemical Name or Material 4-Fluorophenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 207-286-8
RTECSNumber AM0210000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.