Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1765-93-1
C6H6BFO2
139.92
MFCD00039136
LBUNNMJLXWQQBY-UHFFFAOYSA-N
4-fluorobenzeneboronic acid,4-fluorophenyl boronic acid,4-fluorophenyl boranediol,p-fluorophenylboronic acid,4-fluorophenylbornic acid,4-fluorophenyl dihydroxyboron,4-fluorophenyl dihydroxyborane,p-fluorophenyl boric acid,4-fluorophenyl boric acid
285645
CHEBI:48661
(4-fluorophenyl)boronic acid
B(C1=CC=C(C=C1)F)(O)O
1765-93-1
C6H6BFO2
139.92
MFCD00039136
LBUNNMJLXWQQBY-UHFFFAOYSA-N
4-fluorobenzeneboronic acid,4-fluorophenyl boronic acid,4-fluorophenyl boranediol,p-fluorophenylboronic acid,4-fluorophenylbornic acid,4-fluorophenyl dihydroxyboron,4-fluorophenyl dihydroxyborane,p-fluorophenyl boric acid,4-fluorophenyl boric acid
285645
CHEBI:48661
(4-fluorophenyl)boronic acid
B(C1=CC=C(C=C1)F)(O)O
Quantity 1g
Color Yellow to Beige
Packaging Glass bottle
Melting Point 255.0°C to 259.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 139.92
Physical Form Crystalline or Fluffy Powder
Percent Purity 97%
Chemical Name or Material 4-Fluorophenylboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning