Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorothiophenol, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
371-42-6
C6H5FS
128.17
MFCD00004846
OKIHXNKYYGUVTE-UHFFFAOYSA-N
4-fluorothiophenol,p-fluorothiophenol,4-fluorobenzene-1-thiol,4-fluoro thiophenol,p-fluorophenyl mercaptan,benzenethiol, 4-fluoro,4-fluoro-benzenethiol,4-mercaptofluorobenzene,4-fluorthiophenol,p-fluoro-thiophenol
67789
4-fluorobenzenethiol
C1=CC(=CC=C1F)S
371-42-6
C6H5FS
128.17
MFCD00004846
OKIHXNKYYGUVTE-UHFFFAOYSA-N
4-fluorothiophenol,p-fluorothiophenol,4-fluorobenzene-1-thiol,4-fluoro thiophenol,p-fluorophenyl mercaptan,benzenethiol, 4-fluoro,4-fluoro-benzenethiol,4-mercaptofluorobenzene,4-fluorthiophenol,p-fluoro-thiophenol
67789
4-fluorobenzenethiol
C1=CC(=CC=C1F)S
Flash Point 54°C
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5480 to 1.5500
Quantity 10g
Density 54g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H4SH
Formula Weight 128.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Fluorothiophenol
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger