Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Fmoc-amino)butyric acid, 95%

Click to view available options
Quantity:
5g
25g
116821-47-7
C19H19NO4
325.364
MFCD00144889
ACUIFAAXWDLLTR-UHFFFAOYSA-N
fmoc-gaba-oh,fmoc-gamma-abu-oh,fmoc-gaba,n-fmoc-dl-4-amino-butyric acid,4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid,4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid,n-fmoc-4-aminobutyric acid,4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,n-gamma-fmoc-gamma-aminobutyric acid,4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid
2756086
4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O
116821-47-7
C19H19NO4
325.364
MFCD00144889
ACUIFAAXWDLLTR-UHFFFAOYSA-N
fmoc-gaba-oh,fmoc-gamma-abu-oh,fmoc-gaba,n-fmoc-dl-4-amino-butyric acid,4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid,4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid,n-fmoc-4-aminobutyric acid,4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,n-gamma-fmoc-gamma-aminobutyric acid,4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid
2756086
4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O
Applications
4-(Fmoc-amino)butyric acid is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
Quantity 25g
Color White
Linear Formula FmocNHCH2CH2CH2CO2H
Beilstein 7491485
Formula Weight 325.36
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 4-(Fmoc-amino)butyric acid

storageNote1 : 2° to 8°C