Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Formylcinnamic acid, 98%, predominantly trans, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
23359-08-2
C10H8O3
176.17
MFCD00006955
LBOUHDMYVURTMA-AATRIKPKSA-N
4-formylcinnamic acid,3-4-formylphenyl acrylic acid,p-formylcinnamic acid,e-3-4-formylphenyl acrylic acid,2e-3-4-formylphenyl prop-2-enoic acid,4-2-carboxyvinyl benzaldehyde,p-formylcinnamicacid,3-4-formylphenyl prop-2-enoic acid,2e-3-4-formylphenyl acrylic acid,pubchem8244
5357280
(E)-3-(4-formylphenyl)prop-2-enoic acid
C1=CC(=CC=C1C=CC(=O)O)C=O
23359-08-2
C10H8O3
176.17
MFCD00006955
LBOUHDMYVURTMA-AATRIKPKSA-N
4-formylcinnamic acid,3-4-formylphenyl acrylic acid,p-formylcinnamic acid,e-3-4-formylphenyl acrylic acid,2e-3-4-formylphenyl prop-2-enoic acid,4-2-carboxyvinyl benzaldehyde,p-formylcinnamicacid,3-4-formylphenyl prop-2-enoic acid,2e-3-4-formylphenyl acrylic acid,pubchem8244
5357280
(E)-3-(4-formylphenyl)prop-2-enoic acid
C1=CC(=CC=C1C=CC(=O)O)C=O
Packaging Glass bottle
Melting Point 250°C to 255°C
Quantity 1g
Color Yellow to Beige
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HCOC6H4CH=CHCO2H
Formula Weight 176.17
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Formylcinnamic acid, Predominantly trans
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 245-608-9