Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 4 ft. Logic™ Class II, Type A2 Biosafety Cabinet, US Model
SDP

Catalog No. 01812619 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Cabinet
Cabinet, Accessory Package
Cabinet, Accessory Package, Base Stand
Cabinet, Base Stand
Plug Type:
North America (115 V, 15 A)
North America (230 V)
Sash Opening (Metric):
20.3 cm
25 cm
30.5 cm
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Plug Type Sash Opening (Metric)
01-812-619 Cabinet, Base Stand North America (115 V, 15 A) 20.3 cm
01-812-621 Cabinet North America (115 V, 15 A) 20.3 cm
01-812-617 Cabinet, Accessory Package North America (115 V, 15 A) 20.3 cm
01-812-615 Cabinet, Accessory Package, Base Stand North America (115 V, 15 A) 20.3 cm
01-812-637 Cabinet North America (115 V, 15 A) 25 cm
01-812-635 Cabinet, Base Stand North America (115 V, 15 A) 25 cm
01-812-633 Cabinet, Accessory Package North America (115 V, 15 A) 25 cm
01-812-631 Cabinet, Accessory Package, Base Stand North America (115 V, 15 A) 25 cm
01-812-629 Cabinet North America (115 V, 15 A) 30.5 cm
01-812-627 Cabinet, Base Stand North America (115 V, 15 A) 30.5 cm
01-812-625 Cabinet, Accessory Package North America (115 V, 15 A) 30.5 cm
01-812-623 Cabinet, Accessory Package, Base Stand North America (115 V, 15 A) 30.5 cm
01-812-620 Cabinet North America (230 V) 20.3 cm
01-812-618 Cabinet, Base Stand North America (230 V) 20.3 cm
01-812-616 Cabinet, Accessory Package North America (230 V) 20.3 cm
01-812-614 Cabinet, Accessory Package, Base Stand North America (230 V) 20.3 cm
01-812-636 Cabinet North America (230 V) 25 cm
01-812-634 Cabinet, Base Stand North America (230 V) 25 cm
01-812-632 Cabinet, Accessory Package North America (230 V) 25 cm
01-812-630 Cabinet, Accessory Package, Base Stand North America (230 V) 25 cm
01-812-628 Cabinet North America (230 V) 30.5 cm
01-812-626 Cabinet, Base Stand North America (230 V) 30.5 cm
01-812-624 Cabinet, Accessory Package North America (230 V) 30.5 cm
01-812-622 Cabinet, Accessory Package, Base Stand North America (230 V) 30.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 01-812-619 Supplier Labconco™ Supplier No. 302480102
Add to Cart
Add to Cart
Request a Quote

Logic Class II, Type A2 Biosafety Cabinets offer personnel, product and environmental protection from hazardous particulates requiring BSL 1-4 containment. All models ensure the highest standard of safety and are listed to NSF/ANSI 49. Logic cabinets maximize safety, performance, and comfort.

Performance Features:

  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
  • Logic OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
  • Full, six-year warranty on all Class II biosafety cabinets
  • Optional Configurations
  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
  • Telescoping base stand with shelf and leveling feet

Specifications

Description 4 ft. Logic Class II, Type A2 Biosafety Cabinet
Certifications/Compliance NSF/ANSI 49, ETL, ADA, UL, CAN/CSA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel (304)
Width (Metric) Interior 123.2 cm
Product Type Class II, Type A2 Bio Safety Cabinet
Model Logic A2
Noise Level 61 dBA
Outlets 2 Duplex
Plug Type North America (115 V, 15 A)
Power Consumption 180 W
Receptacles 2 Double, Left and Right Side
Height (English) 61.9 in.
Sash Opening (Metric) 20.3 cm
Width (English) Exterior 54.4 in.
Volume (Metric) Exhaust 654 m³/hr.
Display 5 in. touchscreen
Filter Efficiency 99.99%
Filter Type HEPA
Width (Metric) Exterior 138.2 cm
Includes Cabinet, Base Stand
Energy Consumption (Reduced Flow Mode) 37 W
Lighting LED
Height (Metric) 157.2 cm
Sash Opening (English) 8 in.
Service Fixtures 0
Volume 385 CFM
Warranty 6 Years
Width (English) Interior 48.5 in.
Frequency 60 Hz
Height (English) Interior 25.7 in.
Height (English) Max. Opening 22.5 in.
Height (English) Viewing 27 in.
Height (Metric) Interior 65.3 cm
Height (Metric) Max. Opening 57.2 cm
Height (Metric) Viewing 69 cm
Depth (English) 32 in.
Depth (Metric) 81.3 cm
Voltage 115 V
Energy Consumption (Operating Set Point) 180 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.