Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Guanidinobenzoic Acid Methanesulfonate 98.0+%, TCI America™
SDP

Catalog No. g02485g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 148720-07-4
Molecular Formula C9H13N3O5S
Molecular Weight (g/mol) 275.28
MDL Number MFCD00191442
InChI Key NGBBXMQQIAFCGF-UHFFFAOYSA-N
PubChem CID 44630314
IUPAC Name 4-[(diaminomethylidene)amino]benzoic acid; methanesulfonic acid
SMILES CS(O)(=O)=O.NC(N)=NC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 228°C
Color White
Quantity 5 g
Formula Weight 275.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Guanidinobenzoic Acid Methanesulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.