Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptanol 93%, ACROS Organics™

Click to view available options
Quantity:
5mL
Packaging:
Glass bottle
589-55-9
C7H16O
116.2
MFCD00021934
YVBCULSIZWMTFY-UHFFFAOYSA-N
4-heptanol,dipropylcarbinol,unii-yg7b8091bp,di-n-propylcarbinol,heptanol-4,4-heptyl alcohol,n-heptan-4-ol,acmc-209m8k,yvbculsizwmtfy-uhfffaoysa,ch3 ch2 2choh ch2 2ch3
11513
heptan-4-ol
CCCC(CCC)O
589-55-9
C7H16O
116.2
MFCD00021934
YVBCULSIZWMTFY-UHFFFAOYSA-N
4-heptanol,dipropylcarbinol,unii-yg7b8091bp,di-n-propylcarbinol,heptanol-4,4-heptyl alcohol,n-heptan-4-ol,acmc-209m8k,yvbculsizwmtfy-uhfffaoysa,ch3 ch2 2choh ch2 2ch3
11513
heptan-4-ol
CCCC(CCC)O
Melting Point -41.5°C
Flash Point 48°C
Boiling Point 156°C
Specific Gravity 0.81
Density 0.8100g/mL
Refractive Index 1.4175 to 1.4215
Quantity 5mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 91.0
CAS Max % 100.0
Assay Percent Range 93%
Linear Formula CH3(CH2)2CH(OH)C3H7
Solubility Information Solubility in water: immiscible
Formula Weight 116.2
Physical Form Liquid
Percent Purity ≥91%
Chemical Name or Material 4-Heptanol, 93%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning