Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptylaniline 98.0+%, TCI America™
SDP

Catalog No. H058825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H058825ML 25 mL
H05885ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H058825ML Supplier TCI America Supplier No. H058825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37529-27-4
Molecular Formula C13H21N
Molecular Weight (g/mol) 191.32
MDL Number MFCD00007928
InChI Key BNEWZYZRLNNWNR-UHFFFAOYSA-N
Synonym p-heptylaniline, 4-n-heptylaniline, benzenamine, 4-heptyl, 4-heptylphenylamine, acmc-209ito, 4-heptylaniline
PubChem CID 142169
IUPAC Name 4-heptylaniline
SMILES CCCCCCCC1=CC=C(N)C=C1

Specifications

Boiling Point 286°C
Quantity 25 mL
Formula Weight 191.32
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Heptylaniline
EINECSNumber (3)-0206
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.