Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. H07781ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07781ML 1 mL
H07785ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07781ML Supplier TCI America Supplier No. H07781ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60484-67-5
Molecular Formula C14H19N
Molecular Weight (g/mol) 201.313
MDL Number MFCD00143348
InChI Key XTIKBCXMOYZUMG-UHFFFAOYSA-N
Synonym 1-(4-Cyanophenyl)heptane
PubChem CID 575863
IUPAC Name 4-heptylbenzonitrile
SMILES CCCCCCCC1=CC=C(C=C1)C#N

Specifications

Color Yellow
Quantity 1 mL
Formula Weight 201.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Heptylbenzonitrile
EINECSNumber (3)-3541
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.