Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptyn-3-ol, 98%, Thermo Scientific™

Catalog No. AAL1200006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1200006 5 g
AAL1200014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1200006 Supplier Thermo Scientific Chemicals Supplier No. L1200006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32398-69-9
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00041592
InChI Key FMOKAMXPKGORIR-UHFFFAOYSA-N
Synonym 4-heptyn-3-ol, 3-heptyn-5-ol, acmc-1aifi
PubChem CID 141704
IUPAC Name hept-4-yn-3-ol
SMILES CCC#CC(CC)O

Specifications

Boiling Point 94°C to 95°C (54 mmHg)
Flash Point 62°C (143°F)
Quantity 5 g
Beilstein 1699582
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 4-Heptyn-3-ol
Hazard Category Warning
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.