Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexylresorcinol, 98%

Catalog No. AC197921000
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 136-77-6 | C12H18O2 | 194.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-77-6
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00002284
InChI Key WFJIVOKAWHGMBH-UHFFFAOYSA-N
Synonym hexylresorcinol, 4-hexylresorcinol, antascarin, ascaricid, ascarinol, 4-n-hexylresorcinol, oxana, p-hexylresorcinol, adrover, caprokol
PubChem CID 3610
IUPAC Name 4-hexylbenzene-1,3-diol
SMILES CCCCCCC1=CC=C(O)C=C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 65.0°C to 70.0°C
Color Orange to White or Pink
Boiling Point 333.0°C to 335.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 06, II, 904
Merck Index 15, 4748
Solubility Information Solubility in water: 0.5g/l (18°C). Other solubilities: soluble in alcohol,glycerol,ether,chloroform,,acetone and vegetable oils
Formula Weight 194.27
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-Hexylresorcinol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 205-257-4
RTECSNumber VH1575000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.