Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydrazinobenzenesulfonic Acid Hemihydrate 98.0+%, TCI America™
SDP

Catalog No. H0176100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0176100G 100 g
H017625G 25 g
H0176500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0176100G Supplier TCI America Supplier No. H0176100G

Chemical Identifiers

CAS 854689-07-9
Molecular Formula C6H10N2O4S
Molecular Weight (g/mol) 206.216
MDL Number MFCD00480958
InChI Key OSOMMFVBTYFMBI-UHFFFAOYSA-N
Synonym 4-hydrazinylbenzenesulfonic acid hydrate, 4-hydrazinobenzenesulfonic acid hydrate, phenylhydrazine-4-sulfonic acid hemihydrate, benzenesulfonic acid, 4-hydrazino-, monohydrate 9ci
PubChem CID 2849446
IUPAC Name 4-hydrazinylbenzenesulfonic acid;hydrate
SMILES C1=CC(=CC=C1NN)S(=O)(=O)O.O

Specifications

Quantity 100 g
Formula Weight 188.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydrazinobenzenesulfonic Acid Hemihydrate
EINECSNumber (3)-1997
RTECSNumber DB6900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.