Learn More
4-Hydrazinobenzoic acid hydrochloride, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 252.0°C to 254.0°C |
| Assay Percent Range | 96% min. (Argentometry) |
| Packaging | Glass bottle |
| Quantity | 250 g |
| Solubility Information | Solubility in water: soluble. |
| Formula Weight | 188.61 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Hydrazinobenzoic acid hydrochloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
| EINECSNumber | 246-330- |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.