Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine 98.0+%, TCI America™
SDP

Catalog No. H093125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H093125G 25 g
H09315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H093125G Supplier TCI America Supplier No. H093125G

Chemical Identifiers

CAS 52722-86-8
Molecular Formula C11H23NO2
Molecular Weight (g/mol) 201.31
MDL Number MFCD01321174
InChI Key STEYNUVPFMIUOY-UHFFFAOYSA-N
Synonym 1-(2-Hydroxyethyl)-2,2,6,6-tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol
PubChem CID 104301
IUPAC Name 1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidin-4-ol
SMILES CC1(CC(CC(N1CCO)(C)C)O)C

Specifications

Melting Point 186°C
Color White
Quantity 25 g
Formula Weight 201.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.