Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-1-cyclohexanecarboxylic Acid delta-Lactone 98.0+%, TCI America™
SDP

Catalog No. H114325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H114325G 25 g
H11435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H114325G Supplier TCI America Supplier No. H114325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4350-84-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD06797111
InChI Key NVORRKXXHHFWDH-UHFFFAOYSA-N
Synonym 3,6-Ethanotetrahydropyran-2-one, 2-Oxabicyclo[2.2.2]octan-3-one
PubChem CID 138195
IUPAC Name 3-oxabicyclo[2.2.2]octan-2-one
SMILES C1CC2CCC1C(=O)O2

Specifications

Melting Point 128°C
Color White
Boiling Point 123°C
Quantity 25 g
Formula Weight 126.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-1-cyclohexanecarboxylic Acid delta-Lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.