Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-1-indanone 98.0+%, TCI America™
SDP

Catalog No. H072325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H072325G 25 g
H07235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H072325G Supplier TCI America Supplier No. H072325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40731-98-4
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00143330
InChI Key CKSCMRNFDBWFND-UHFFFAOYSA-N
Synonym 4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659
PubChem CID 590547
IUPAC Name 4-hydroxy-2,3-dihydro-1H-inden-1-one
SMILES OC1=CC=CC2=C1CCC2=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 148.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-1-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.