Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy, 98%, free radical, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2226-96-2
C9H18NO2
172.25
MFCD00006478
4-Hydroxy-TEMPO
2226-96-2
C9H18NO2
172.25
MFCD00006478
4-Hydroxy-TEMPO
Polymerization reagent
Viscosity 20 mPa.s (40°C)
Infrared Spectrum Authentic
Packaging Glass bottle
Color Orange
Quantity 25g
Melting Point 69.0°C to 73.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Fieser 09,247
Merck Index 15, 9284
Solubility Information Solubility in water: soluble.
Formula Weight 172.25
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy, Free radical
Show More Show Less

GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if swallowed.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 218-760-9