Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-2-butanone 95.0+%, TCI America™
SDP

Catalog No. H0584100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0584100ML 100 mL
H058425ML 25 mL
H0584500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0584100ML Supplier TCI America Supplier No. H0584100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 590-90-9
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00059005
InChI Key LVSQXDHWDCMMRJ-UHFFFAOYSA-N
Synonym 4-hydroxy-2-butanone, 2-butanone, 4-hydroxy, methylolacetone, 3-oxo-1-butanol, 3-oxobutanol, 3-ketobutan-1-ol, monomethylolacetone, 4-butanol-2-one, unii-tcm0bj44mf, 2-hydroxyethyl methyl ketone
PubChem CID 111509
ChEBI CHEBI:41268
IUPAC Name 4-hydroxybutan-2-one
SMILES CC(=O)CCO

Specifications

Color Colorless
Boiling Point 170°C
Quantity 100 mL
Formula Weight 88.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-2-butanone
EINECSNumber (2)-3210
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.