Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Hydroxy-2-hexenal Antibody (5C11.1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318263
This item is not returnable.
View return policy
Description
4-Hydroxy-2-hexenal Monoclonal antibody specifically detects 4-Hydroxy-2-hexenal in All species samples. It is validated for Western Blot, ELISA, Dot BlotSpecifications
4-Hydroxy-2-hexenal | |
Monoclonal | |
1 mg/ml | |
Western Blot 1:1000, ELISA 1:1000, Dot Blot | |
Mouse | |
Protein G purified | |
RUO | |
All species | |
Purified |
Western Blot, ELISA, Dot Blot | |
5C11.1 | |
Unconjugated | |
PBS pH 7.4, 50% glycerol | |
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH) | |
100 μg | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction