Learn More
4-Hydroxy-2-hexenal Antibody (6F10), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259352G
Catalog No. NBP259352G
Only null left
Add to Cart
Description
Description
4-Hydroxy-2-hexenal Monoclonal specifically detects 4-Hydroxy-2-hexenal in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
4-Hydroxy-2-hexenal
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for 4-Hydroxy-2-hexenal (4-HHE) and 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxy-2-hexenal (4-HHE) or 4-Hydroxynonenal (4-HNE). Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins.
Store at 4C in the dark.
IgG2b
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
6F10
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Mouse
Protein G purified
Primary
Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.