Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Hydroxy-2-hexenal Antibody (6F10), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259352MFV450
Description
4-Hydroxy-2-hexenal Monoclonal antibody specifically detects 4-Hydroxy-2-hexenal in Non-species specific samples. It is validated for Flow CytometrySpecifications
4-Hydroxy-2-hexenal | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Non-species specific | |
Purified |
Flow Cytometry | |
6F10 | |
Flow Cytometry | |
4-HHE;4-hydroxy Hexenal;4-Hydroxy-2-hexenal;4-Hydroxy-2-hexenal-modified;HHE;MG;MG-modified protein | |
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction