Learn More
Invitrogen™ 4-Hydroxy-2-hexenal Monoclonal Antibody (5C11.1), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545860
Description
A 1:1,000 dilution of MA5-45860 was sufficient for detection of 4-Hydroxy-2-hexenal in 1 μg of 4-Hydroxy-2-hexenal-BSA conjugate by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Specific for 4-Hydroxy-2-hexenal. Slight cross-reactivity with 4-Hydroxynonenal. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins.
4-Hydroxynonenal, or 4-hydroxy-2-nonenal or 4-HNE or HNE, (C9H16O2), is an alpha,beta-unsaturated hydroxyalkenal that is produced by lipid peroxidation in cells. 4-HNE is the primary alpha,beta-unsaturated hydroxyalkenal formed in this process.
Specifications
4-Hydroxy-2-hexenal | |
Monoclonal | |
1 mg/mL | |
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
Dot Blot, ELISA, Western Blot | |
5C11.1 | |
PE | |
4-HHE; 4-hydroxy Hexenal; 4-Hydroxy-2-hexenal; 4-Hydroxy-2-hexenal-modified; HHE; MG; MG-modified protein | |
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH). | |
100 μg | |
Primary | |
4°C | |
Liquid | |
Chemical |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.