Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 4-Hydroxy-2-hexenal Monoclonal Antibody (5C11.1), PerCP
GREENER_CHOICE

Catalog No. PIMA545859
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545859 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA545859 Supplier Invitrogen™ Supplier No. MA545859
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45859 was sufficient for detection of 4-Hydroxy-2-hexenal in 1 μg of 4-Hydroxy-2-hexenal-BSA conjugate by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Specific for 4-Hydroxy-2-hexenal. Slight cross-reactivity with 4-Hydroxynonenal. Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins.

4-Hydroxynonenal, or 4-hydroxy-2-nonenal or 4-HNE or HNE, (C9H16O2), is an alpha,beta-unsaturated hydroxyalkenal that is produced by lipid peroxidation in cells. 4-HNE is the primary alpha,beta-unsaturated hydroxyalkenal formed in this process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-Hydroxy-2-hexenal
Applications Dot Blot, ELISA, Western Blot
Classification Monoclonal
Clone 5C11.1
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene Alias 4-HHE; 4-hydroxy Hexenal; 4-Hydroxy-2-hexenal; 4-Hydroxy-2-hexenal-modified; HHE; MG; MG-modified protein
Host Species Mouse
Immunogen Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.