Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methoxy-DL-mandelic acid, 98%

Catalog No. AAA1334906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1334906 5 g
AAA1334903 1 g
AAA1334914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1334906 Supplier Thermo Scientific Chemicals Supplier No. A1334906
Add to Cart

CAS: 55-10-7 | C9H10O5 | 198.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-10-7
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00004235
InChI Key CGQCWMIAEPEHNQ-UHFFFAOYSA-N
Synonym vanillylmandelic acid, vanilmandelic acid, dl-4-hydroxy-3-methoxymandelic acid, 2-hydroxy-2-4-hydroxy-3-methoxyphenyl acetic acid, 4-hydroxy-3-methoxymandelic acid, vanillomandelic acid, +/--vanillylmandelic acid, vma, alpha,4-dihydroxy-3-methoxybenzeneacetic acid, benzeneacetic acid, .alpha.,4-dihydroxy-3-methoxy
PubChem CID 1245
ChEBI CHEBI:20106
IUPAC Name 2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)C(C(=O)O)O)O

Specifications

Melting Point 133°C to 134°C (decomposition)
Quantity 5 g
Formula Weight 198.17
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-methoxy-DL-mandelic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-224-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.