Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methoxymandelic acid, 99%

Catalog No. AC164900050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164900050 5 g Glass bottle
AC164900010 1 g Glass bottle
AC164900250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164900050 Supplier Thermo Scientific Chemicals Supplier No. 164900050
Add to Cart

CAS: 55-10-7 | C9H9O5 | 197.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-10-7
Molecular Formula C9H9O5
Molecular Weight (g/mol) 197.17
MDL Number MFCD00004235
InChI Key CGQCWMIAEPEHNQ-QMMMGPOBSA-M
Synonym vanillylmandelic acid, vanilmandelic acid, dl-4-hydroxy-3-methoxymandelic acid, 2-hydroxy-2-4-hydroxy-3-methoxyphenyl acetic acid, 4-hydroxy-3-methoxymandelic acid, vanillomandelic acid, +/--vanillylmandelic acid, vma, alpha,4-dihydroxy-3-methoxybenzeneacetic acid, benzeneacetic acid, .alpha.,4-dihydroxy-3-methoxy
PubChem CID 1245
ChEBI CHEBI:20106
SMILES COC1=CC(=CC=C1O)[C@H](O)C([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 131.0°C to 134.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CH(OH)CO2H
Quantity 5 g
Merck Index 15,10121
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in acetone,moderatly soluble in ether and acetonitrile,sparingly soluble in benzene
IUPAC Name (2S)-2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetate
Formula Weight 198.17
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxymandelic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-224-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.