Learn More
4-Hydroxy-3-methyl-2-butanone, tech 85%
Description
Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo vinylation and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Protonated diacetone alcohol and protonated 4- hydroxy-3-methyl-2-butanone undergo “vinylation” and colli- sion-induced dehydration as the only primary reactions, the product of the former reaction consisting of up to 40% of the total product ion distribution.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Refractive Index | 1.4325 |
| Quantity | 1 g |
| Formula Weight | 102.13 |
| Percent Purity | 85% |
| Grade | Technical |
| Chemical Name or Material | 4-Hydroxy-3-methyl-2-butanone |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 222-238-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.