Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methylbenzaldehyde, 98%

Catalog No. ac427550010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

CAS: 15174-69-3 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15174-69-3
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00012360
InChI Key BAKYASSDAXQKKY-UHFFFAOYSA-N
Synonym 4-hydroxy-3-methyl-benzaldehyde, benzaldehyde, 4-hydroxy-3-methyl, unii-p8cux6265j, 3-methyl-4-hydroxybenzaldehyde, pubchem8134, acmc-1buur, 6-hydroxy-m-tolualdehyde, intermediates-zcf02161, ksc494o2r, 3-methyl-p-hydroxybenzaldehyde
PubChem CID 139901
IUPAC Name 4-hydroxy-3-methylbenzaldehyde
SMILES CC1=CC(C=O)=CC=C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 118°C to 120°C
Color Brown to Tan-White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(OH)CHO
Quantity 1 g
Formula Weight 136.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methylbenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.