Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-4-methyl-2-pentanone, 99+%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
123-42-2
C6H12O2
116.16
SWXVUIWOUIDPGS-UHFFFAOYSA-N
diacetone alcohol,4-hydroxy-4-methyl-2-pentanone,diacetonalkohol,diketone alcohol,diacetonalcohol,tyranton,acetonyldimethylcarbinol,diacetone-alcool,pyranton a,2-methyl-2-pentanol-4-one
31256
CHEBI:55381
4-hydroxy-4-methylpentan-2-one
CC(=O)CC(C)(C)O
123-42-2
C6H12O2
116.16
SWXVUIWOUIDPGS-UHFFFAOYSA-N
diacetone alcohol,4-hydroxy-4-methyl-2-pentanone,diacetonalkohol,diketone alcohol,diacetonalcohol,tyranton,acetonyldimethylcarbinol,diacetone-alcool,pyranton a,2-methyl-2-pentanol-4-one
31256
CHEBI:55381
4-hydroxy-4-methylpentan-2-one
CC(=O)CC(C)(C)O
Viscosity 3.22 mPa.s (20°C)
Color Colorless
Specific Gravity 0.9385
Melting Point -42.8°C
Packaging Glass bottle
Refractive Index 1.4223 to 1.4243
Quantity 1L
Flash Point 58°C
Boiling Point 166.0°C
Density 0.9385g/mL
Assay Percent Range 99%
Linear Formula (CH3)2C(OH)CH2COCH3
Merck Index 15,2967
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,ether
Formula Weight 116.16
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material 4-Hydroxy-4-methyl-2-pentanone
Show More Show Less

EINECSNumber : 204-626-7