Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzophenone, 98%

Catalog No. aaa1509618
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 1137-42-4 | C13H10O2 | 198.22 g/mol

4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-42-4
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002355
InChI Key NPFYZDNDJHZQKY-UHFFFAOYSA-N
Synonym 4-hydroxybenzophenone, 4-benzoylphenol, 4-hydroxyphenyl phenyl methanone, p-benzoylphenol, p-hydroxybenzophenone, methanone, 4-hydroxyphenyl phenyl, 4-hydroxyphenyl phenylmethanone, benzophenone, 4-hydroxy, unii-04r2lws0ms, 4'-hydroxybenzophenone
PubChem CID 14347
ChEBI CHEBI:34421
IUPAC Name (4-hydroxyphenyl)-phenylmethanone
SMILES OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 132°C to 135°C
Boiling Point 260°C to 262°C (24 mmHg)
Quantity 50 g
Beilstein 777776
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-507-1
RTECSNumber PC4959775
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.