Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzyl alcohol, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
623-05-2
C7H8O2
124.14
BVJSUAQZOZWCKN-UHFFFAOYSA-N
4-hydroxybenzyl alcohol,4-hydroxymethyl phenol,p-hydroxybenzyl alcohol,p-methylolphenol,4-methylolphenol,4-hydroxybenzenemethanol,benzenemethanol, 4-hydroxy,p-hydroxymethyl phenol,gastrodigenin,4-hydroxybenzylalcohol
125
CHEBI:67410
4-(hydroxymethyl)phenol
C1=CC(=CC=C1CO)O
623-05-2
C7H8O2
124.14
BVJSUAQZOZWCKN-UHFFFAOYSA-N
4-hydroxybenzyl alcohol,4-hydroxymethyl phenol,p-hydroxybenzyl alcohol,p-methylolphenol,4-methylolphenol,4-hydroxybenzenemethanol,benzenemethanol, 4-hydroxy,p-hydroxymethyl phenol,gastrodigenin,4-hydroxybenzylalcohol
125
CHEBI:67410
4-(hydroxymethyl)phenol
C1=CC(=CC=C1CO)O
Melting Point 110.0°C to 112.0°C
Color Pink to Beige
Quantity 500g
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula HOC6H4CH2OH
Beilstein 06,897
Solubility Information Solubility in water: soluble
Formula Weight 124.14
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzyl alcohol
Show More Show Less