Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzylideneacetone, 97%

Catalog No. aaa1760514
Click to view available options
Quantity:
25 g
100 g

CAS: 3160-35-8 | C10H10O2 | 162.19 g/mol

4-Hydroxybenzylideneacetone is used in the synthesis of 4-(4-hydroxy-phenyl)-butan-2-one. This reaction needs the reagents of cyclohexane and AlCl3, and the solvent of CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzylideneacetone is used in the synthesis of 4-(4-hydroxy-phenyl)-butan-2-one. This reaction needs the reagents of cyclohexane and AlCl3, and the solvent of CH2Cl2.

Solubility
Optimally soluble in aqueous solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3160-35-8
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00016490
InChI Key OCNIKEFATSKIBE-NSCUHMNNSA-N
Synonym p-hydroxybenzalacetone, 4-hydroxybenzylideneacetone, 4-hydroxybenzal acetone, 4-hydroxycinnamoylmethane, 4-hydroxybenzalacetone, p-hydroxybenzylidene acetone, 4-p-hydroxyphenyl-3-buten-2-one, 4-4-hydroxyphenyl but-3-en-2-one, 3-buten-2-one, 4-4-hydroxyphenyl, 3-buten-2-one, 4-p-hydroxyphenyl
PubChem CID 796857
SMILES CC(=O)\C=C\C1=CC=C(O)C=C1

Specifications

Melting Point 106°C to 111°C
Quantity 25 g
Beilstein 2412544
Solubility Information Optimally soluble in aqueous solvents.
IUPAC Name (E)-4-(4-hydroxyphenyl)but-3-en-2-one
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzylideneacetone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-607-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.