Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymercury)benzoic Acid Sodium Salt, 98%

Catalog No. p-145438
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 138-85-2 | C7H5HgNaO3 | 360.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-85-2
Molecular Formula C7H5HgNaO3
Molecular Weight (g/mol) 360.69
MDL Number MFCD00002527
InChI Key BERBQUNVJGVZCX-UHFFFAOYSA-M
Synonym sodium 4-carboxylatophenyl mercury hydrate, sodium hydrate p-mercuribenzoate
PubChem CID 23667696
IUPAC Name sodium;(4-carboxylatophenyl)mercury;hydrate
SMILES C1=CC(=CC=C1C(=O)[O-])[Hg].O.[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Hg)
Packaging Glass bottle
Quantity 1 g
Beilstein 16, 969
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 360.69
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Hydroxymercury)benzoic acid sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Fatal in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-340-5
RTECSNumber OV8150000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.