Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxymethyl-5-methylimidazole 96.0+%, TCI America™
SDP

Catalog No. H097625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H097625G 25 g
H09765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H097625G Supplier TCI America Supplier No. H097625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29636-87-1
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.13
MDL Number MFCD00792476
InChI Key AXJZCJSXNZZMDU-UHFFFAOYSA-N
PubChem CID 122433
IUPAC Name (5-methyl-1H-imidazol-4-yl)methanol
SMILES CC1=C(CO)N=CN1

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 25 g
Formula Weight 112.13
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxymethyl-5-methylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.