Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymethyl)imidazole hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
32673-41-9
C4H6N2O·ClH
134.57
MFCD00012697
WFNASTYGEKUMIY-UHFFFAOYSA-N
4-imidazolemethanol hydrochloride,4-hydroxymethyl imidazole hydrochloride,1h-imidazol-4-yl methanol hydrochloride,4-hydroxymethylimidazole hydrochloride,imidazol-4-ylmethanol hydrochloride,1h-imidazol-4-ylmethanol hydrochloride,1h-imidazol-5-ylmethanol hydrochloride,4-hydroxymethyl imidazole hcl,1h-imidazole-4-ylmethanol hydrochloride,4 5-hydroxymethylimidazole hydrochloride
122926
1H-imidazol-5-ylmethanol;hydrochloride
C1=C(NC=N1)CO.Cl
32673-41-9
C4H6N2O·ClH
134.57
MFCD00012697
WFNASTYGEKUMIY-UHFFFAOYSA-N
4-imidazolemethanol hydrochloride,4-hydroxymethyl imidazole hydrochloride,1h-imidazol-4-yl methanol hydrochloride,4-hydroxymethylimidazole hydrochloride,imidazol-4-ylmethanol hydrochloride,1h-imidazol-4-ylmethanol hydrochloride,1h-imidazol-5-ylmethanol hydrochloride,4-hydroxymethyl imidazole hcl,1h-imidazole-4-ylmethanol hydrochloride,4 5-hydroxymethylimidazole hydrochloride
122926
1H-imidazol-5-ylmethanol;hydrochloride
C1=C(NC=N1)CO.Cl
Melting Point 110°C to 112°C
Infrared Spectrum Authentic
Color Beige to Yellow or Brown
Packaging Glass bottle
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 23, I, 103
Formula Weight 134.57
Physical Form Crystalline Powder and/Chunks
Percent Purity 99%
Chemical Name or Material 4-(Hydroxymethyl)imidazole hydrochloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 251-150-