Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymethyl)imidazole hydrochloride, 99%, Thermo Scientific™

Catalog No. AC204870250
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 32673-41-9
Molecular Formula C4H7ClN2O
Molecular Weight (g/mol) 134.56
MDL Number MFCD00012697
InChI Key WFNASTYGEKUMIY-UHFFFAOYSA-N
Synonym 4-imidazolemethanol hydrochloride, 4-hydroxymethyl imidazole hydrochloride, 1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethylimidazole hydrochloride, imidazol-4-ylmethanol hydrochloride, 1h-imidazol-4-ylmethanol hydrochloride, 1h-imidazol-5-ylmethanol hydrochloride, 4-hydroxymethyl imidazole hcl, 1h-imidazole-4-ylmethanol hydrochloride, 4 5-hydroxymethylimidazole hydrochloride
PubChem CID 122926
IUPAC Name 1H-imidazol-5-ylmethanol;hydrochloride
SMILES Cl.OCC1=CN=CN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 110°C to 112°C
Color Beige to Yellow or Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Beilstein 23, I, 103
Formula Weight 134.57
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-(Hydroxymethyl)imidazole hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 251-150-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.