Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193335 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. H1204-5G

Supplier: TCI America H12045G

Only null left
Add to Cart
Add to Cart

Specifications

CAS 59016-93-2
Color White-Yellow
Molecular Formula C7H9BO3
MDL Number MFCD00792672
Quantity 5 g
Synonym 4-hydroxymethyl phenylboronic acid, 4-hydroxymethyl benzeneboronic acid, 4-hydroxymethyl phenyl boronic acid, 4-hydroxymethylphenyl boronic acid, 4-hydroxymethylphenylboronic acid
InChI Key PZRPBPMLSSNFOM-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)CO)(O)O
IUPAC Name [4-(hydroxymethyl)phenyl]boronic acid
Molecular Weight (g/mol) 151.956
PubChem CID 2734706
Formula Weight 151.96
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.