Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymethyl)tetrahydropyran, 98%

Catalog No. p-7041691
Click to view available options
Quantity:
1 g
5 g

CAS: 14774-37-9 | C6H12O2 | 116.16 g/mol

Used as a reagent in the identification and optimization of pteridinone Toll-like receptor 7 agonists, which can be used for the oral treatment of viral hepatitis. 4-(Hydroxymethyl)tetrahydropyran is also a starting material for [1-[(Tetrahydro-2H-pyran-4-yl)methyl]-1H-indol-3-yl](2,2,3,3-tetramethylcyclopropyl)methanone which is a drug that acts as CB2 cannabinoid receptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a reagent in the identification and optimization of pteridinone Toll-like receptor 7 agonists, which can be used for the oral treatment of viral hepatitis. 4-(Hydroxymethyl)tetrahydropyran is also a starting material for [1-[(Tetrahydro-2H-pyran-4-yl)methyl]-1H-indol-3-yl](2,2,3,3-tetramethylcyclopropyl)methanone which is a drug that acts as CB2 cannabinoid receptor agonist.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14774-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00457804
InChI Key YSNVSVCWTBLLRW-UHFFFAOYSA-N
Synonym tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol
PubChem CID 2773573
SMILES OCC1CCOCC1

Specifications

Boiling Point 111°C to 115°C (15 mmHg)
Flash Point 61°C (142°F)
Refractive Index 1.46
Quantity 1 g
Beilstein 103069
Solubility Information Slightly soluble in water.
IUPAC Name oxan-4-ylmethanol
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 4-(Hydroxymethyl)tetrahydropyran
Hazard Category H227-H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.