Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxynonenal Mouse, HRP, Clone:12F7, Abnova™

Mouse monoclonal antibody raised against synthetic 4-Hydroxynonenal (4-HNE).

Supplier:  Abnova Corporation MAB17402

Catalog No. 89-334-270


Only null left
Add to Cart

Description

Description

Specifications

Specifications

4-Hydroxynonenal
Monoclonal
HRP
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Synthetic 4-Hydroxynonenal modified Keyhole Limpet Hemocyanin (KLH).
Store at -20°C.
Liquid
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
12F7
ELISA (1:1000)
Immunocytochemistry (1:50)
Immunofluorescence (1:50)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Mouse
100 μg
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.