Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193920 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 4-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 71597-85-8
Melting Point 232°C
Molecular Formula C6H7BO3
Quantity 1 g
MDL Number MFCD01074628
Synonym 4-hydroxyphenyl boronic acid, 4-hydroxybenzeneboronic acid, 4-boronophenol, 4-hydroxyphenyl boranediol, p-hydroxyphenylboronic acid, 4-hydroxyphenyl-boronic acid, boronic acid, 4-hydroxyphenyl, pubchem1729, acmc-209ojb
InChI Key COIQUVGFTILYGA-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(O)C=C1
IUPAC Name (4-hydroxyphenyl)boronic acid
Molecular Weight (g/mol) 137.93
PubChem CID 2734360
Formula Weight 137.93
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.