Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyphenyldimethylsulfonium Methyl Sulfate 98.0+%, TCI America™
SDP

Catalog No. H102225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H102225G 25 g
H10225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H102225G Supplier TCI America Supplier No. H102225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32279-04-2
Molecular Formula C9H14O5S2
Molecular Weight (g/mol) 266.326
InChI Key PKQZVASULXKBJV-UHFFFAOYSA-N
Synonym Dimethyl-4-hydroxyphenylsulfonium Methyl Sulfate
PubChem CID 21915988
IUPAC Name (4-hydroxyphenyl)-dimethylsulfanium;methyl sulfate
SMILES COS(=O)(=O)[O-].C[S+](C)C1=CC=C(C=C1)O

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 25 g
Formula Weight 266.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyphenyldimethylsulfonium Methyl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.