Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyquinoline-2-carboxylic acid, hydrate, 98%

Catalog No. AC121910250
Click to view available options
Quantity:
5 g
25 g

Catalog No. AC121910250

Supplier: Thermo Scientific Chemicals 121910250

Only null left
Add to Cart

CAS: 345909-35-5 | C10H7NO3 | 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345909-35-5
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00149476
InChI Key HCZHHEIFKROPDY-UHFFFAOYSA-N
Synonym 4-hydroxyquinoline-2-carboxylic acid hydrate, kynurenic acid hydrate, kynurenic acid monohydrate, 4-hydroxyquinaldic acid hydrate, c10h7no3.h2o, 2-carboxy-4-hydroxyquinoline hydrate, 4-hydroxyquinoline-2-carboxylic acid, hydrate, 4-hydroxyquinoline-2-carboxylic acid,hydrate,98, 4-hydroxy-2-quinolinecarboxylic acid hydrate 1:1
PubChem CID 18530299
IUPAC Name 4-oxo-1H-quinoline-2-carboxylic acid;hydrate
SMILES OC(=O)C1=NC2=CC=CC=C2C(O)=C1

Specifications

Melting Point 275.0°C
Infrared Spectrum Authentic
Quantity 25 g
Beilstein 22,23
Merck Index 14,5327
Solubility Information Solubility in water: 0.9g/L water (100°C). Other solubilities: soluble in hot alcohol, insoluble in ether, very soluble in alkali
Formula Weight 189.17
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinoline-2-carboxylic acid, hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.