Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxythiobenzamide 98.0+%, TCI America™
SDP

Catalog No. H130825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H130825G 25 g
H13085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H130825G Supplier TCI America Supplier No. H130825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25984-63-8
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.20
MDL Number MFCD04973332
InChI Key DOCQBKMMNPJLOR-UHFFFAOYSA-N
Synonym 4-hydroxythiobenzamide, 4-hydroxybenzothioamide, 4-hydroxy thiobenzamide, 4-hydroxybenzene-1-carbothioamide, 4-hydroxybenzenecarbothioamide, benzenecarbothioamide, 4-hydroxy, 4-hydroxy-thiobenzamide, 4-aminothioxomethyl phenol, 4-amino sulfanyl methylidene cyclohexa-2,5-dien-1-one, p-hydroxythiobenzamide
PubChem CID 5706487
IUPAC Name 4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES NC(S)=C1C=CC(=O)C=C1

Specifications

Melting Point 192°C
Color Yellow
Quantity 25 g
Formula Weight 153.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxythiobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.