Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-3-(trifluoromethyl)-1-H-pyrazole, 97%, Thermo Scientific™

Catalog No. AC444820010
Click to view available options
:
1g; Glass bottle

Specifications

CAS 866638-72-4
Melting Point 139.0°C to 141.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C4H2F3IN2
Quantity 1 g
Synonym 4-iodo-3-trifluoromethyl-1h-pyrazole, 4-iodo-5-trifluoromethyl-1h-pyrazole, 1h-pyrazole, 4-iodo-3-trifluoromethyl, acmc-209qaw, 4-iodo-3-trifluoromethyl pyrazole, 3-trifluoromethyl-4-iodo-1h-pyrazole, 4-iodo-3-trifluoromethyl-2h-pyrazole
InChI Key XRUHSOXIZMYOHX-UHFFFAOYSA-N
SMILES C1=NNC(=C1I)C(F)(F)F
IUPAC Name 4-iodo-5-(trifluoromethyl)-1H-pyrazole
Molecular Weight (g/mol) 261.97
PubChem CID 16069792
Formula Weight 261.97
Percent Purity 97%
Chemical Name or Material 4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.