Learn More
4-Iodo-L-phenylalanine, 95%
Description
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.
Solubility
Partly miscible with water.
Notes
Light sensitive. Incompatible with oxidizing agents.

Specifications
Specifications
Beilstein | 4411317 |
Solubility Information | Partly miscible with water. |
Quantity | 5 g |
Formula Weight | 291.09 |
Optical Rotation | −6.5° (c=1 in Acetic Acid:Water (4:1)) |
Percent Purity | 95% |
Sensitivity | Light sensitive |
Chemical Name or Material | 4-Iodo-L-phenylalanine |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.