Alfa Aesar™ 4-Iodobenzenesulfonyl chloride, 97%

Manufacturer: Alfa Aesar™ B2441622

Quantity 100g

 View more versions of this product

Catalog No. AAB2441622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98-61-3
    Molecular Formula C6H4ClIO2S
    Formula Weight 302.52
    MDL Number MFCD00007441
    UN Number UN3261
    Beilstein 2691661
    Merck Index 14,7486
    Synonym Pipsyl chloride
    Chemical Name or Material 4-Iodobenzenesulfonyl chloride

    Specifications

    Formula Weight 302.52
    MDL Number MFCD00007441
    UN Number UN3261
    Beilstein 2691661
    Merck Index 14,7486
    Quantity 100g
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    TSCA TSCA
    Molecular Formula C6H4ClIO2S
    CAS 98-61-3
    Melting Point 79° to 84°C
    Synonym Pipsyl chloride
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material 4-Iodobenzenesulfonyl chloride
    Assay 97%

    Description

    Applications
    It is used as intermediate in organic syntheses & in medicine.

    Solubility
    Reacts with water.

    Notes
    Moisture & light sensitive. Store away from oxidizing agent.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit