Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodopyrazole 98.0+%, TCI America™
SDP

Catalog No. I069225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I069225G 25 g
I06925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I069225G Supplier TCI America Supplier No. I069225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3469-69-0
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.975
MDL Number MFCD00005244
InChI Key LLNQWPTUJJYTTE-UHFFFAOYSA-N
Synonym 4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295
PubChem CID 77022
ChEBI CHEBI:45260
IUPAC Name 4-iodo-1H-pyrazole
SMILES C1=C(C=NN1)I

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 25 g
Formula Weight 193.98
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodopyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.