Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropoxyaniline 98.0+%, TCI America™
SDP

Catalog No. I0399100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0399100G 100 g
I03995G 5 g
I039925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0399100G Supplier TCI America Supplier No. I0399100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-66-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00059188
InChI Key MLNFMFAMNBGAQT-UHFFFAOYSA-N
PubChem CID 82112
IUPAC Name 4-propan-2-yloxyaniline
SMILES CC(C)OC1=CC=C(C=C1)N

Specifications

Color Yellow
Boiling Point 95°C
Quantity 100 g
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Isopropoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.