Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. I08065G
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 153624-46-5
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD03427051
InChI Key CJUHQADBFQRIMC-UHFFFAOYSA-N
Synonym 4-isopropoxyphenylboronic acid, 4-isopropoxylphenylboronic acid, 4-isopropoxyphenyl boronic acid, 4-isopropoxybenzeneboronic acid, 4-isopropyloxyphenylboronic acid, 4-propan-2-yloxyphenyl boronic acid, boronic acid, 4-1-methylethoxy phenyl, 4-propan-2-yloxy phenyl boronic acid, 4-iso-propoxyphenylboronic acid, p-isopropoxyphenylboronic acid
PubChem CID 3698726
IUPAC Name (4-propan-2-yloxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OC(C)C)(O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 180.01
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.